missing translation for 'onlineSavingsMsg'
Learn More

Barium metaborate monohydrate, Thermo Scientific™

Product Code. 15492457
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492457 5 kg 5kg
15472457 250 g 250g
15482457 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492457 Supplier Thermo Scientific Alfa Aesar Supplier No. 045823.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-06-9
Molecular Formula B2BaH2O5
Molecular Weight (g/mol) 240.96
MDL Number MFCD00046182
InChI Key JXOXZJCHHKIXMI-UHFFFAOYSA-N
Synonym barium metaborate monohydrate, 2bo2.ba.h2o, barium 2+ ;oxido oxo borane;hydrate, barium 2+ hydrate dioxoborinate, barium 2+ ion hydrate dioxoborinate
PubChem CID 23165673
SMILES O.[Ba++].[O-]B=O.[O-]B=O

Specifications

Melting Point >900°C
Color White
Physical Form Crystalline
Quantity 5 kg
Linear Formula BaB2O4·H2O
UN Number UN1564
Solubility Information Soluble in water. Moderately soluble in dilute acids. Insoluble in organic solvents.
IUPAC Name barium(2+);oxido(oxo)borane;hydrate
Formula Weight 240.97 (222.96 Anhydrous)
Density 3.35 g/mL
Chemical Name or Material Barium metaborate monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.