missing translation for 'onlineSavingsMsg'
Learn More

Barium manganate(VI), 95%, Thermo Scientific Chemicals

Product Code. 10741345 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10741345 5 g Glass bottle 5g
10348480 25 g Glass bottle 25g
10422011 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10741345 Supplier Thermo Scientific Acros Supplier No. 198480050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-35-1
Molecular Formula BaMnO4
Molecular Weight (g/mol) 256.26
MDL Number MFCD00011514
InChI Key ZZCNKSMCIZCVDR-UHFFFAOYSA-N
Synonym barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese
PubChem CID 3084030
IUPAC Name barium(2+);dioxido(dioxo)manganese
SMILES [Ba++].[O-][Mn]([O-])(=O)=O

Specifications

Color Black-Blue to Blue
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 94% min
Linear Formula BaMnO4
Fieser 08,21; 09,23; 10,16; 12,38; 15,18; 16,18
Merck Index 15, 977
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acids
Formula Weight 256.27
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Barium manganate(VI), 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium manganate(VI)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.