missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium isopropoxide, 20% w/v in isopropanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mL
50 mL
Description
Barium isopropoxide is used in nano-materials and thin Films, catalysis synthesis, ferroelectric metals. It is also used as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Density | 0.89 g/mL |
Flash Point | 22°C (72°F) |
Odor | Alcohol-like |
Linear Formula | Ba[OCH(CH3)2]2 |
Quantity | 10 mL |
UN Number | UN3274 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in alcohols,ketones and esters. Reacts with water. |
Formula Weight | 255.52 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'