missing translation for 'onlineSavingsMsg'
Learn More

Barium isopropoxide, 20% w/v in isopropanol

Product Code. 11344048
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344048 50 mL 50mL
11354048 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344048 Supplier Thermo Scientific Alfa Aesar Supplier No. 041338.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

In Sol-Gel synthesis and superconductors | CAS: 24363-37-9 | C6H14BaO2 | 255.503 g/mol

Barium isopropoxide is used in nano-materials and thin Films, catalysis synthesis, ferroelectric metals. It is also used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 24363-37-9
Molecular Formula C6H14BaO2
Molecular Weight (g/mol) 255.503
MDL Number MFCD00050486
InChI Key CPUJSIVIXCTVEI-UHFFFAOYSA-N
Synonym barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate
PubChem CID 6101105
IUPAC Name barium(2+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].[Ba+2]

Specifikationer

Color Yellow
Density 0.89 g/mL
Flash Point 22°C (72°F)
Odor Alcohol-like
Linear Formula Ba[OCH(CH3)2]2
Quantity 50 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohols,ketones and esters. Reacts with water.
Formula Weight 255.52
Concentration or Composition (by Analyte or Components) 20% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Barium isopropoxide
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.