missing translation for 'onlineSavingsMsg'
Learn More

Barium isopropoxide, Thermo Scientific Chemicals

Product Code. 11332478
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332478 5 g 5g
11352478 1 g 1g
11342478 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332478 Supplier Thermo Scientific Alfa Aesar Supplier No. 014523.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24363-37-9
Molecular Formula C6H14BaO2
Molecular Weight (g/mol) 255.503
MDL Number MFCD00050486
InChI Key CPUJSIVIXCTVEI-UHFFFAOYSA-N
Synonym barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate
PubChem CID 6101105
IUPAC Name barium(2+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].[Ba+2]

Specifications

Melting Point ∼200°C (decomposition)
Color Brown to Yellow
Physical Form Powder
Quantity 5 g
Linear Formula Ba[OCH(CH3)2]2
UN Number UN3205
Formula Weight 255.52
Sensitivity Moisture Sensitive
Chemical Name or Material Barium isopropoxide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.