missing translation for 'onlineSavingsMsg'
Learn More

Barium iodide, ultra dry, 99.995% (metals basis)

Product Code. 11333847
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333847 5 g 5g
11343847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333847 Supplier Thermo Scientific Alfa Aesar Supplier No. 035679.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light Sensitive and Hygroscopic | CAS: 13718-50-8 | BaI2 | 391.14 g/mol

Barium iodide is used to prepare organobarium compounds by reacting with alkyl potassium compounds. It is also involved in the preparation of active barium metal by reducing with lithium biphenyl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-50-8
Molecular Formula BaI2
Molecular Weight (g/mol) 391.14
MDL Number MFCD00003452
InChI Key SGUXGJPBTNFBAD-UHFFFAOYSA-L
IUPAC Name barium(2+) diiodide
SMILES [I-].[I-].[Ba++]

Specifications

Melting Point 740°C
Color White
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 391.15
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.15
Chemical Name or Material Barium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.