missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Hydroxide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Barium hydroxide |
| CAS | 17194-00-2 |
| Molecular Formula | Ba H2 O2 |
| Quantity | 25 g |
| Synonym | Barium Dihydroxide, Caustic Baryta |
| InChI Formula | InChI=1S/Ba.2H2O/h;2*1H2/q+2;;/p-2 |
| SMILES | O[Ba]O |
| IUPAC Name | dihydroxybarium |
| Molecular Weight (g/mol) | 171.34 |
| Formula Weight | 171.9107 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction