missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Octahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10283560 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10283560 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10283560 Supplier Fisher Chemical Supplier No. B/0650/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
IUPAC Name barium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78°C
Color White
pH 12.5
Physical Form Solid
Quantity 1 kg
Formula Weight 315.48g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.