missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, 98+%, for analysis

Product Code. 10083523 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083523 5 kg Plastic bottle 5kg
10286830 250 g Plastic bottle 250g
10646422 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10083523 Supplier Thermo Scientific Acros Supplier No. 203140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium hydroxide octahydrate, 98+%, for analysis, Quantity: 1kg, Packaging: Plastic bottle, CAS: 12230-71-6, White, Grade: Analysis, Melting Point: 78.0 deg.C, Molecular Formula: H2BaO2.8H2O, Molecular Weight: 315.48, Percent Purity: 98+% | CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78.0°C
Color White
Physical Form Crystals, Crystalline Flakes or Powder
Quantity 5 kg
Assay Percent Range 98+%
Linear Formula Ba(OH)2·8H2O
Merck Index 15, 973
Solubility Information Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol,slightly soluble in ethanol,practically insoluble in acetone
IUPAC Name barium(2+);dihydroxide;octahydrate
Formula Weight 315.48
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.18
Density 2.1800g/mL
Chemical Name or Material Barium hydroxide octahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.