missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, 97%

Product Code. 11450643
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450643 10,000 g 10000g
11430643 500 g 500g
11440643 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450643 Supplier Thermo Scientific Alfa Aesar Supplier No. A12714.0C
  • ONLINE EXCLUSIVE PRICE
    £260.00
    £234.65 / 10000g
    Save £25.35
    9% Off
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

Alfa Aesar Barium hydroxide octahydrate, 97%, Quantity: 500g, CAS: 12230-71-6, Melting Point: 78 deg.C, Molecular Formula: BaH18O10, Molecular Weight: 315.461, Percent Purity: 97% | CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium hydroxide octahydrate has been used in the centromeric heterochromatin banding technnique and to synthesize BaTiO3. The form Barium hydroxide was reported to catalyze the rate of β elimination of phosphoserine residues. It is also used as a general purpose additive for lubricants and greases, sugar fabrication, manufacturing soaps, fat saponification and chemical synthesis of other barium compounds.

Solubility
Soluble in water (72 mg/ml at 20°C), methanol, ethanol (slightly), and water (56 mg/ml at 15°C). Insoluble in acetone (practically)

Notes
Air Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from air and oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78°C
Quantity 10,000 g
Linear Formula Ba(OH)2·8H2O
UN Number UN3262
Merck Index 14,977
Solubility Information Soluble in water (72mg/ml at 20°C),methanol,ethanol (slightly),and water (56mg/ml at 15°C). Insoluble in acetone (practically)
IUPAC Name barium(2+);dihydroxide;octahydrate
Formula Weight 315.48 (171.36 Anhydrous)
Percent Purity 97%
Odor Odorless
Sensitivity Air Sensitive
Density 2.18
Chemical Name or Material Barium hydroxide octahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium hydroxide octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Barium hydroxide octahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.