missing translation for 'onlineSavingsMsg'
Learn More

Barium Hydroxide Monohydrate, purum p.a., ≥98.0% (T), Honeywell™

Product Code. 15621080 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15621080 100 g Poly Bottle 100g
15631080 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15621080 Supplier Honeywell Research Chemicals Supplier No. 11783100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22326-55-2
Molecular Formula BaH4O3
Molecular Weight (g/mol) 189.356
MDL Number MFCD00149151
InChI Key GKQTUHKAQKWLIN-UHFFFAOYSA-L
Synonym barium hydroxide monohydrate, barium hydroxide ba oh 2 , monohydrate, bariumdiol hydrate, barium hydroxide hydrate trace metals basis, barium hydroxide monohydrate, purum p.a t
PubChem CID 16211219
IUPAC Name barium(2+);dihydroxide;hydrate
SMILES O.[OH-].[OH-].[Ba+2]

Specifications

Boiling Point 780°C
Color White
pH 13.1 (21.0g/L at 25°C)
Quantity 100 g
Linear Formula Ba(OH)2 · H2O
UN Number UN3262
Formula Weight 189.36g/mol
Percent Purity ≥98.0% (T)
Grade purum p.a.
Packaging Poly Bottle
Appearance Powder
Vapor Pressure 300 hPa at 78°C
Density 3.743g/mL at 25°C
Chemical Name or Material Barium Hydroxide Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.