missing translation for 'onlineSavingsMsg'
Learn More

Barium fluoride, 99%

Product Code. p-4781805
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 11331709

Brand: Thermo Scientific Alfa Aesar 012338.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

It finds applications in ceramic flux, glass manufacturing, metallurgy, electronics and dry-film lubricants. It is transparent from ultraviolet to infrared. Barium fluoride is usually employed as a scintillator material, and useful as a transmission window material for IR spectroscopy, mostly used for analysis of fuel oil samples. Of all the optical fluorides, it is the most resistant to high-energy radiation. It is used as a preopacifying agent, and in the production of enamel, welding agents, and glazing frits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Color Colorless
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Soluble in ethanol,methanol,and aqueous solutions of ammonium chloride. Slightly soluble in water.
IUPAC Name barium(2+) difluoride
Formula Weight 175.36
Odor Odorless
Refractive Index 1.4741
Density 4.83 g/cm3
Chemical Name or Material Barium fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.