missing translation for 'onlineSavingsMsg'
Learn More

Barium fluoride, 99.9% (metals basis)

Product Code. 11377828
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377828 100 g 100g
11367828 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377828 Supplier Thermo Scientific Alfa Aesar Supplier No. 036380.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

Barium fluoride is used as a material to make optical components such as lenses, in windows for infrared spectroscopy, in particular in the field of fuel oil analysis, used as a preopacifying agent and in enamel and glazing frits production. It is also used in the production of welding agents, metallurgy, as a molten bath for refining aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Color Colorless
Physical Form Fused Lump
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in methanol,ethanol,water 1.2g/L.
IUPAC Name barium(2+);difluoride
Formula Weight 175.36
Odor Odorless
Refractive Index 1.4741
Density 4.83
Chemical Name or Material Barium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.