missing translation for 'onlineSavingsMsg'
Learn More

Barium Fluoride, 98%, Honeywell™ Fluka™

Product Code. 15652330 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15652330 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15652330 Supplier Honeywell-Fluka Supplier No. 236101500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2260°C
Melting Point 1350°C
CAS 7787-32-8
Color White
Physical Form Powder
Quantity 500 g
Molecular Formula BaF2
MDL Number MFCD00003450
UN Number UN1564
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES [F-].[F-].[Ba++]
IUPAC Name barium(2+);difluoride
Molecular Weight (g/mol) 175.32
PubChem CID 62670
Formula Weight 175.32g/mol
Percent Purity 98%
Packaging Poly bottle
Density 4.89g/mL at 25°C
Chemical Name or Material Barium Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.