missing translation for 'onlineSavingsMsg'
Learn More

Barium ethoxide, 99.5% (metals basis), 10% w/v in ethanol

Product Code. 11328028
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328028 25 mL 25mL
11338028 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328028 Supplier Thermo Scientific Alfa Aesar Supplier No. 036588.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2914-19-4 | C4H10BaO2 | 227.449 g/mol

It is used in nano-materials & thin films; catalysis & synthesis and optics & glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2914-19-4
Molecular Formula C4H10BaO2
Molecular Weight (g/mol) 227.449
MDL Number MFCD00050488
InChI Key GYIWFHXWLCXGQO-UHFFFAOYSA-N
Synonym barium diethanolate, diethoxybarium, barium 2+ ; ethanolate, ethanol, barium salt 9ci, barium 2+ bis ethoxide, barium 2+ ion bis ethoxide, barium ethoxide solution w/v in ethanol trace metals basis 25ml
PubChem CID 6451474
IUPAC Name barium(2+);ethanolate
SMILES CC[O-].CC[O-].[Ba+2]

Specifications

Color Yellow
Linear Formula Ba(OC2H5)2
Quantity 25 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 227.42
Percent Purity 99.5%
Concentration or Composition (by Analyte or Components) 10% w/v in ethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Barium ethoxide
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.