missing translation for 'onlineSavingsMsg'
Learn More

Barium dodecairon nonadecaoxide, Thermo Scientific Chemicals

Catalog Number 15422357
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
12047-11-9
BaFe12O19
MFCD00075637
Barium ferrite
This item is not returnable. View return policy
12047-11-9
BaFe12O19
MFCD00075637
Barium ferrite

Barium dodecairon nonadecaoxide is used in subwoofer magnets to provide power. It is also used in new high tech magnetic tapes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >1315°C
Color Black
Physical Form Powder
Quantity 25 g
UN Number UN1564
Solubility Information Insoluble in water.
Formula Weight 1111.48
Chemical Name or Material Barium dodecairon nonadecaoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'