missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Barium diphenylaminesulfonate, pure, indicator grade

Product Code. 10658144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10658144 50 g Glass bottle 50g
10045730 5 g Glass bottle 5g
10443351 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10658144 Supplier Thermo Scientific Chemicals Supplier No. 401680500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6211-24-1 | C24H20BaN2O6S2 | 633.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6211-24-1
Molecular Formula C24H20BaN2O6S2
Molecular Weight (g/mol) 633.88
MDL Number MFCD00007497
InChI Key IVCNVXFNTKXMCA-UHFFFAOYSA-L
Synonym barium diphenylamine-4-sulfonate, barium diphenylaminesulfonate, benzenesulfonic acid, 4-phenylamino-, barium salt 2:1, barium bis 4-anilinobenzenesulphonate, barium diphenylamine sulfonate, n-phenylsufanilic acid barium, unii-ssj5f79159, diphenylamine-4-sulfonic acid barium salt, n-phenylsufanilic acid, barium salt, 4-anilinobenzenesulfonate; barium 2+
PubChem CID 80342
IUPAC Name 4-anilinobenzenesulfonate;barium(2+)
SMILES [Ba++].[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1.[O-]S(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 300.0°C
Color White to Gray
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 50 g
Sensitivity (as indicator) passes
Merck Index 15, 7423
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,insoluble in ether and benzene
Formula Weight 633.89
Grade Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Barium diphenylaminesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Barium diphenylaminesulfonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.