missing translation for 'onlineSavingsMsg'
Learn More

Barium chromate, 98%

Product Code. 11346978
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
missing translation for 'unitSize'
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346978 1 kg 1kg
11356978 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11346978 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 014669.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-40-3 | BaCrO4 | 253.32 g/mol

Barium chromate is used as a pigment, oxidizing agent, colorant in glass, and sulfate scavenger in chromium electroplating baths, ceramics, and porcelain. It acts as a corrosion inhibitor in jointing pastes and metal primers. Further, it is used in fuses, safety matches, ignition control devices and high-temperature batteries. In addition to this, it is used as a carrier for the chromium ions and useful as a burn rate modifier in pyrotechnic compositions. It is involved as a catalyst in the alkane dehydration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-40-3
Molecular Formula BaCrO4
Molecular Weight (g/mol) 253.32
MDL Number MFCD00014180
InChI Key QFFVPLLCYGOFPU-UHFFFAOYSA-N
Synonym barium chromate, lemon chrome, permanent yellow, barium chromate vi, baryta yellow, ultramarine yellow, pigment yellow 31, ci pigment ylllow 31, barium chromate 1:1, unii-7d7o9cf0ix
PubChem CID 25136
SMILES [Ba++].[O-][Cr]([O-])(=O)=O

Specifications

Color Yellow
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98%
Solubility Information Insoluble in water,acetic acid and chromic acid.
IUPAC Name barium(2+) dioxochromiumbis(olate)
Formula Weight 253.33
Odor Odorless
Density 4.5
Chemical Name or Material Barium chromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.