missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium chromate, Puratronic™, 99.995% (metals basis)

Brand:  Alfa Aesar™ 011124.06

Code : NEW

Additional Details : CAS Number : 10294-40-3 Weight : 0.00500kg Transport : UN number : 1479 Chem class : 5.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11306148

  • £43.20 / 5g


Estimated Shipment date
from Supplier 16-07-2018
Add to basket

Specifications

Chemical Identifiers

Barium chromate
Puratronic™
MFCD00014180
14,972
10294-40-3
253.33
UN1479

Specifications

Barium chromate
Puratronic™
99.998%
253.33
UN1479
4.50g/mL
Yellow
10294-40-3
Powder
(metals basis)
MFCD00014180
14,972
Odorless
5g
Videos
SDS