missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, ultra dry, 99.95% (metals basis), Thermo Scientific™

Catalog Number 11332498
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10361-37-2
BaCl2
208.23
MFCD00003445
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
25204
CHEBI:63317
[Cl-].[Cl-].[Ba++]
This item is not returnable. View return policy
10361-37-2
BaCl2
208.23
MFCD00003445
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
25204
CHEBI:63317
[Cl-].[Cl-].[Ba++]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1560°C
Melting Point 963°C
Color White
Physical Form Beads
Quantity 5 g
UN Number UN1564
Merck Index 14,971
Solubility Information Soluble in water 370g/L.
IUPAC Name barium(2+) dichloride
Formula Weight 208.25
Percent Purity 99.95%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.9 g/mL
Assay (metals basis)
Chemical Name or Material Barium chloride, Ultra dry
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'