Learn More
Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
£56.10 - £653.00
Chemical Identifiers
CAS | 10326-27-9 |
---|---|
Molecular Formula | BaCl2H4O2 |
Molecular Weight (g/mol) | 244.26 |
MDL Number | MFCD00149154 |
InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
PubChem CID | 5284346 |
ChEBI | CHEBI:86153 |
IUPAC Name | barium(2+) dihydrate dichloride |
SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11310849
|
Thermo Scientific Alfa Aesar
010646.14 |
25 g |
£56.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330849
|
Thermo Scientific Alfa Aesar
010646.22 |
100 g |
£176.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320849
|
Thermo Scientific Alfa Aesar
010646.36 |
500 g |
£653.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
barium(2+) dihydrate dichloride |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 | |
O.O.[Cl-].[Cl-].[Ba++] |
Specifications
113°C-2H2O | |
White | |
99.99% (Metal basis) | |
MFCD00149154 | |
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate. | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
Odorless | |
3.097 g/cm3 |
10326-27-9 | |
Powder | |
BaCl2H4O2 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
5284346 | |
244.28 (208.25 Anhydrous) | |
White powder | |
Barium chloride dihydrate, dihydrate |
Safety and Handling
P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
H301-H319-H332
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 600-412-6
missing translation for 'rtecsNumber' : CQ8751000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only