missing translation for 'onlineSavingsMsg'
Learn More

Barium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31750490
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
Unit Size:
1000g
100g
2.50 L
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31750490 100 g White Poly Wide Mouth Bottle 100g
31750209 500 g White Poly Wide Mouth Bottle 500g
31750701 1 kg White Poly Wide Mouth Bottle 1000g
31749965 2.5 kg Natural Poly Bottle 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31750490 Supplier Ricca Chemical Company Supplier No. RDCB0080100B1
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Physical Form Crystals or Powder
Quantity 100 g
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
Percent Purity ≥99%
Grade ACS
Loss on Drying 14.0 to 16.0%
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Barium Chloride, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.