missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, 99+%, for analysis

Product Code. p-3734649 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10276830

Brand: Thermo Scientific Acros 203135000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560.0°C
Melting Point 962.0°C
Color Gray-White
pH 5.2 to 8.2 (5% soln. at 25°C)
Quantity 500 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula BaCl2·2H2O
Merck Index 15, 967
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, practically insoluble in ethanol, acetone and, ethyl acetate
Formula Weight 244.28
Percent Purity ≥99%
Grade Analytical
Loss on Drying 14.0 % to 16.0 % (150°C)
Packaging Plastic bottle
Chemical Name or Material Barium chloride dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Barium chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.