missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

£40.80 - £131.00

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10158040
View Documents Promotion Details
Thermo Scientific Acros
423455000
500 g Plastic bottle
£40.80
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10298843
View Documents Promotion Details
Thermo Scientific Acros
423450025
2.5 kg Plastic bottle
£131.00
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10326-27-9
244.26
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
barium(2+) dihydrate dichloride
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
CHEBI:86153
O.O.[Cl-].[Cl-].[Ba++]

Specifications

1560.0°C
10326-27-9
5.2 to 8.2 (5% soln. at 25°C)
99% min. (Complexometry)
BaCl2·2H2O
15, 967
Solubility in water: soluble. Other solubilities: soluble in methanol, practically insoluble in ethanol, acetone and, ethyl acetate
O.O.[Cl-].[Cl-].[Ba++]
244.26
CHEBI:86153
≥99%
14.0% to 16.0% (150°C)
962.0°C
White
Crystals
BaCl2H4O2
MFCD00149154
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium(2+) dihydrate dichloride
5284346
244.28
ACS Reagent
Barium chloride dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
If eye irritation per

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : CQ8751000

Product Identifier
  • Barium chloride dihydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4

Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only