missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, anhydrous, 99.998% (metals basis)

Product Code. p-7048601
Click to view available options
Quantity:
0.5 g
5 g
25 g
100 g
Unit Size:
0.5g
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11351598

Brand: Thermo Scientific Alfa Aesar 010995.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10361-37-2 | BaCl2 | 208.23 g/mol

Barium chloride is used in manufacture of pigments, fire works, other barium salts, fireworks , hardening of steel, heat treatment salts, purification of brine solution in caustic chlorine plants, lubrication oil additive, textile dye, pigments, white leather, aluminum refining, boiling water treatment and porcelain enamels for sheet steel. It is also used to remove sulfate ion in some electrolytic plants, caustic soda, magnesium metal or sodium metal. It is also used as a component in a flux used to prevent oxidation of molten magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 963°C
Color White
Physical Form Lump/Powder
Quantity 0.5 g
UN Number UN1564
Merck Index 14,971
Solubility Information Soluble in water and methanol. Insoluble in acids,ethanol,acetone and ethyl acetate. Slightly soluble in nitric acid and hydrochloric acid.
IUPAC Name barium(2+);dichloride
Formula Weight 208.25
Percent Purity 99.998%
Odor Odorless
Sensitivity Hygroscopic
Density 3.9 g/mL
Assay (metals basis)
Chemical Name or Material Barium chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.