missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium chlorate monohydrate, 98%, Thermo Scientific™

Product Code. 11439382
Change view
Click to view available options
Quantity:
2500 g
Unit Size:
2500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439382 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11439382 Supplier Alfa Aesar™ Supplier No. A10128.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-38-9
Molecular Formula BaCl2H2O7
Molecular Weight (g/mol) 322.236
MDL Number MFCD00149147
InChI Key CIADYQPMTWAHHH-UHFFFAOYSA-L
Synonym barium chlorate monohydrate, barium 2+ dichlorate hydrate, ba.2clo3.h2o, chloric acid, bariumsalt 2:1, barium 2+ hydrate dichlorate
PubChem CID 6096944
IUPAC Name barium(2+);dichlorate;hydrate
SMILES O.[O-]Cl(=O)=O.[O-]Cl(=O)=O.[Ba+2]

Specifications

Melting Point 414°C
Quantity 2500 g
UN Number UN1445
Formula Weight 322.26 (304.24 Anhydrous)
Percent Purity 98%
Density 3.18
Chemical Name or Material Barium chlorate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.