missing translation for 'onlineSavingsMsg'
Learn More

Barium chloranilate trihydrate

Product Code. 10239550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10239550 25 g Plastic bottle 25g
10538512 100 g Plastic bottle 100g
11954881 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10239550 Supplier Thermo Scientific Acros Supplier No. 401660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32458-20-1 | C6H8BaCl2O7 | 400.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32458-20-1
Molecular Formula C6H8BaCl2O7
Molecular Weight (g/mol) 400.35
MDL Number MFCD00078327
InChI Key JFBLKYVZQCFPJE-UHFFFAOYSA-N
Synonym barium chloranilate, reag
PubChem CID 131877766
IUPAC Name barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
SMILES O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O

Specifications

Color Brown
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 398.34
Physical Form Powder
Chemical Name or Material Barium chloranilate trihydrate
compliance-icons
Product Identifier
  • Barium chloranilate trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.