missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, Puriss. p.a., ACS reagent, Honeywell Fluka™

Product Code. 16675733 Shop All Honeywell Fluka Products
Change view
Click to view available options
:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16675733 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16675733 Supplier Honeywell Fluka Supplier No. 1172925KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 513-77-9
Color White
CAS Max % ≤ 100%
pH 7 to 8
Quantity 25 kg
Molecular Formula CBaO3
Linear Formula BaCO3
MDL Number MFCD00003448
Beilstein 7045119
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES [Ba++].[O-]C([O-])=O
IUPAC Name barium(2+) carbonate
Molecular Weight (g/mol) 197.34
Grade ACS
Density 4.4 g/cm3
Chemical Name or Material Barium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.