missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, Puratronic™, 99.997% (metals basis)

Product Code. 11390839
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390839 500 g 500g
11300849 25 g 25g
11380839 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11390839 Supplier Thermo Scientific Alfa Aesar Supplier No. 010645.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 811°C
Physical Form Lumps, Flakes and Granular Powder
Quantity 500 g
Merck Index 14,969
Solubility Information Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid.
IUPAC Name barium(2+);carbonate
Formula Weight 197.35
Percent Purity 99.997%
Grade Puratronic
Odor Odorless
Density 4.43 g/mL
Assay (metals basis)
Chemical Name or Material Barium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.