missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number 11380839
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O
This item is not returnable. View return policy
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O

Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point (decomposition)
Melting Point 811°C
Physical Form Lumps, Flakes and Granular Powder
Quantity 100 g
Merck Index 14,969
Solubility Information Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid.
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 4.43 g/mL
Assay (metals basis)
Chemical Name or Material Barium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'