Learn More
Barium carbonate, ACS, 99.0-101.0%, Thermo Scientific Chemicals
Description
Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | (decomposition) |
Melting Point | 811°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.0 to 101.0% |
Solubility Information | Soluble in most acids,with the exception of sulfuric acid. It is insoluble in water and ethanol. |
IUPAC Name | barium(2+) carbonate |
Formula Weight | 197.35 |
Grade | ACS |
Show More |
RUO – Research Use Only