missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, 99%

Product Code. 11424614
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
1000g
100g
5000g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424614 1 kg 1000g
11374599 100 g 100g
11384599 500 g 500g
11434614 5 kg 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11424614 Supplier Thermo Scientific Alfa Aesar Supplier No. A13195.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

Barium carbonate is widely used in the ceramics industry as an ingredient in glazes. It acts as a flux, a matting and crystallizing agent and combines with certain colouring oxides to produce unique colours not easily attainable by other means. It is also used for removing of sulfates in phosphoric acid production and chlorine alkali electrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Barium carbonate is widely used in the ceramics industry as an ingredient in glazes. It acts as a flux, a matting and crystallizing agent and combines with certain colouring oxides to produce unique colours not easily attainable by other means. It is also used for removing of sulfates in phosphoric acid production and chlorine alkali electrolysis.

Solubility
Insoluble in water and alcohol. Soluble in acetic acid, hydrochloric acid and nitric acid.

Notes
m.p. 174° (90 atm), 811° (1 atm)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Melting Point 811°C
Quantity 1 kg
Linear Formula BaCO3
Merck Index 14,969
Solubility Information Insoluble aq. soln. and alcohol. Soluble in acetic acid,hydrochloric acid and nitric acid.
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Percent Purity 99%
Odor Odorless
Density 4.43
Chemical Name or Material Barium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Barium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : Barium carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.