missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, 99+%, for analysis

Product Code. 10217363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217363 2.5 kg Plastic bottle 2.5kg
10412221 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217363 Supplier Thermo Scientific Acros Supplier No. 217490025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-77-9 | CBaO3 | 197.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point 1450.0°C
Melting Point 811.0°C
Color White
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula BaCO3
Merck Index 15, 965
Solubility Information Solubility in water: 0.002g/100mL (20°C)
IUPAC Name barium(2+);carbonate
Formula Weight 197.35
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 4.43
Density 4.4300g/mL
Chemical Name or Material Barium carbonate, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.