missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1450.0°C |
| Melting Point | 811.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 99+% |
| Linear Formula | BaCO3 |
| Merck Index | 15, 965 |
| Solubility Information | Solubility in water: 0.002g/100mL (20°C) |
| IUPAC Name | barium(2+) carbonate |
| Show More |
Safety and Handling
Product Identifier
- Barium carbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Barium carbonate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction