missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium calcium copper oxide (2-1-2), 99.9% (metals basis)

Brand:  Alfa Aesar™ 039192.09

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 1564 Chem class : 6.1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11393568

  • £45.10 / 10g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Barium calcium copper oxide (2-1-2)
MFCD00211652
UN1564

Specifications

Barium calcium copper oxide (2-1-2)
99.9%
MFCD00211652
10g
Fine Agglomerate & Ballmilled Powder
(metals basis)
UN1564
Videos
SDS