missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific™

£25.92 - £303.00

Chemical Identifiers

CAS 7791-28-8
Molecular Formula BaBr2H4O2
Molecular Weight (g/mol) 333.17
MDL Number MFCD00149153
InChI Key HQQMRSGBIPFNSN-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dibromide
SMILES O.O.[Br-].[Br-].[Ba++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326148
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011125.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11336148
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011125.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11346148
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011125.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7791-28-8
333.17
HQQMRSGBIPFNSN-UHFFFAOYSA-L
O.O.[Br-].[Br-].[Ba++]
BaBr2H4O2
MFCD00149153
barium(2+) dihydrate dibromide

Specifications

7791-28-8
Powder
MFCD00149153
14,968
HQQMRSGBIPFNSN-UHFFFAOYSA-L
barium(2+) dihydrate dibromide
333.17 (297.14 Anhydrous)
Puratronic™
3.58 g/mL
Barium bromide dihydrate
White
BaBr2H4O2
UN1564
Very soluble in water, soluble in methanol
O.O.[Br-].[Br-].[Ba++]
333.17
99.999%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 234-140-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications