missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium bromide dihydrate, 99.3%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Barium bromide dihydrate is used to purify radium and make zinc bromide. It also used precursor to chemicals used in photograph.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 99.30% |
Solubility Information | Very soluble in water,soluble in methanol |
Formula Weight | 333.17 (297.14 Anhydrous) |
Density | 3.58 g/mL |
Chemical Name or Material | Barium bromide dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'