missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide dihydrate, 98+%

Product Code. 15473137
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473137 500 g 500g
15463137 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473137 Supplier Thermo Scientific Alfa Aesar Supplier No. A16328.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7791-28-8 | BaBr2H4O2 | 333.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-28-8
Molecular Formula BaBr2H4O2
Molecular Weight (g/mol) 333.17
MDL Number MFCD00149153
InChI Key HQQMRSGBIPFNSN-UHFFFAOYSA-L
IUPAC Name barium(2+) dihydrate dibromide
SMILES O.O.[Br-].[Br-].[Ba++]

Specifications

Quantity 500 g
UN Number UN1564
Merck Index 14,968
Formula Weight 333.17 (297.15 Anhydrous)
Percent Purity ≥98%
Sensitivity Hygroscopic
Density 3.58
Chemical Name or Material Barium bromide dihydrate
compliance-icons
Product Identifier
  • Barium bromide dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Barium bromide dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.