missing translation for 'onlineSavingsMsg'
Learn More

Barium bromate, 97%

Product Code. p-7081159
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11444473

Brand: Thermo Scientific Alfa Aesar A11008.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13967-90-3 | BaBr2O6 | 393.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13967-90-3
Molecular Formula BaBr2O6
Molecular Weight (g/mol) 393.129
MDL Number MFCD00014177
InChI Key VEASZGAADGZARC-UHFFFAOYSA-L
Synonym barium bromate, bromic acid, barium salt, unii-5qk4064y0r, barium 2+ dibromate, acmc-1bumj, ba.2bro3, bromic acid, barium salt 2:1, barium bromate un2719 oxidizer
PubChem CID 61706
IUPAC Name barium(2+);dibromate
SMILES [O-]Br(=O)=O.[O-]Br(=O)=O.[Ba+2]

Specifications

Quantity 100 g
UN Number UN2719
Formula Weight 393.16
Percent Purity 97%
Odor Odorless
Density 3.99
Chemical Name or Material Barium bromate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.