missing translation for 'onlineSavingsMsg'
Learn More

Barium bromate, 97%

Product Code. 11454473
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454473 500 g 500g
11444473 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454473 Supplier Thermo Scientific Alfa Aesar Supplier No. A11008.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13967-90-3 | BaBr2O6 | 393.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13967-90-3
Molecular Formula BaBr2O6
Molecular Weight (g/mol) 393.129
MDL Number MFCD00014177
InChI Key VEASZGAADGZARC-UHFFFAOYSA-L
Synonym barium bromate, bromic acid, barium salt, unii-5qk4064y0r, barium 2+ dibromate, acmc-1bumj, ba.2bro3, bromic acid, barium salt 2:1, barium bromate un2719 oxidizer
PubChem CID 61706
IUPAC Name barium(2+);dibromate
SMILES [O-]Br(=O)=O.[O-]Br(=O)=O.[Ba+2]

Specifications

Quantity 500 g
UN Number UN2719
Formula Weight 393.16
Percent Purity 97%
Odor Odorless
Density 3.99
Chemical Name or Material Barium bromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.