missing translation for 'onlineSavingsMsg'
Learn More

Barium bis(trifluoromethylsulfonyl)imide, Thermo Scientific™

Product Code. 15452677
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168106-22-7
Molecular Formula C4H2BaF12N2O8S4
Molecular Weight (g/mol) 699.614
MDL Number MFCD22200529
InChI Key VJUKAFJLZPVWME-UHFFFAOYSA-N
Synonym barium bis trifluoromethanesulfonimide, barium bis-triflimide
PubChem CID 131725281
IUPAC Name barium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Ba]

Specifications

Melting Point >300°C
Color White to Yellow
Linear Formula [(CF3SO2)2N]2Ba
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Formula Weight 697.6
Physical Form Solid
Chemical Name or Material Barium bis (trifluoromethylsulfonyl)imide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'