missing translation for 'onlineSavingsMsg'
Learn More

Barium aluminum oxide, tech.

Product Code. 11398067
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398067 10 g 10g
11308077 100 g 100g
11318077 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398067 Supplier Thermo Scientific Alfa Aesar Supplier No. 022642.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12004-04-5

Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Frary’s metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-04-5
MDL Number MFCD00049716
Synonym Barium aluminate

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN1564
Solubility Information Insoluble in water.
Formula Weight 561.98
Grade Technical
Odor Odorless
Chemical Name or Material Barium aluminum oxide
compliance-icons
Product Identifier
  • Barium aluminum oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.