missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium aluminum oxide, tech., Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
Description
Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Frary’s metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 500 g |
UN Number | UN1564 |
Solubility Information | Insoluble in water. |
Formula Weight | 561.98 |
Grade | Technical |
Odor | Odorless |
Chemical Name or Material | Barium aluminum oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'