missing translation for 'onlineSavingsMsg'
Learn More

Barium Acetate, TRC

Product Code. 30334933
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334933 5 g 5g
30332251 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30334933 Supplier Toronto Research Chemicals Supplier No. TRCB1186855G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium Acetate
CAS 543-80-6
Molecular Formula Ba. 2(C2H4O2)
Quantity 5 g
Synonym Barium(II) Acetate, Barium Diacetate
InChI Formula InChI=1S/2C2H4O2.Ba/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2
SMILES [Ba+2].CC(=O)[O-].CC(=O)[O-]
IUPAC Name barium(2+);diacetate
Molecular Weight (g/mol) 255.41
Formula Weight 255.9318
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.