missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, Puratronic™, 99.999% (metals basis)

Product Code. 11376148
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376148 25 g 25g
11366148 5 g 5g
11356148 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376148 Supplier Thermo Scientific Alfa Aesar Supplier No. 011126.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Linear Formula Ba(OOCCH3)2
UN Number UN1564
Beilstein 3693411
Merck Index 14,965
IUPAC Name barium(2+) diacetate
Formula Weight 255.43
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 2.47 g/mL
Assay (metals basis)
Chemical Name or Material Barium acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.