missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, 99+%, ACS reagent

Product Code. 10608994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10608994 2.5 kg Plastic bottle 2.5kg
10583494 100 g Plastic bottle 100g
10197890 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10608994 Supplier Thermo Scientific Acros Supplier No. 423430025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Tekniske data

Color White
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 99.0 to 102% (Complexometry)
Linear Formula (CH3CO2)2Ba
Merck Index 15, 961
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 255.43
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.46
Density 2.4600g/mL
Chemical Name or Material Barium acetate
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Barium acetate
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
Faresætning
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.