missing translation for 'onlineSavingsMsg'
Learn More

Barium AA Standard: Ba @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17958979
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
17958979 500 mL 500mL
17968649 100 mL 100mL
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17958979

Brand: VHG Labs VHGAABAN500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Barium AA Standard: Ba @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium
CAS 7440-39-3
Methods AA/GFAA
Analyte or Component Names Barium
Shipping Condition Room Temperature
Molecular Formula Ba
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Bario, Baryum
InChI Formula InChI=1S/Ba
SMILES [Ba]
IUPAC Name barium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 137.33
Quantity 500 mL
Formula Weight 137.905 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.