missing translation for 'onlineSavingsMsg'
Learn More

Barium 2-ethylhexanoate, 20% w/w in 2-ethylhexanoic acid, Thermo Scientific™

£29.40 - £430.00

Chemical Identifiers

CAS 2457-01-4
MDL Number MFCD00058696
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355098
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044021.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11365098
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044021.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11345098
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044021.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2457-01-4
MFCD00058696

Specifications

2457-01-4
C16H30O4Sr
strontium 2-ethylhexanoate, strontium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, strontium salt 2:1, acmc-20ajmv, bis 2-ethylhexanoic acid strontium salt, hexanoic acid, 2-ethyl-, strontium salt, strontium 2+ bis 2-ethylhexanoate, hexanoic acid,2-ethyl-, strontium salt 2:1
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sr+2]
374.03
423.76
Liquid
Weak, characteristic
MFCD00058696
DLWBHRIWCMOQKI-UHFFFAOYNA-L
strontium;2-ethylhexanoate
102822
20% w/w in 2-ethylhexanoic acid
Barium 2-ethylhexanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H332
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P281-P308+P313-P501c

H361d

missing translation for 'einecsNumber' : 219-535-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications