missing translation for 'onlineSavingsMsg'
Learn More

Barium 2,4-pentanedionate

Product Code. p-7049533
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11382568

Brand: Thermo Scientific Alfa Aesar 014613.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12084-29-6 | C10H14BaO4 | 335.55 g/mol

Barium 2,4-pentanedionate hydrate is used in the preparation of radioluminescence of poly ethylene glycolated Eu3+ doped nanophosphors as bioimaging probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12084-29-6
Molecular Formula C10H14BaO4
Molecular Weight (g/mol) 335.55
MDL Number MFCD00013484
InChI Key DJHZYHWLGNJISM-FDGPNNRMSA-L
Synonym barium 2+ ; z-4-oxopent-2-en-2-olate, barium 2,4-pentanedionate, bis pentane-2,4-dionato-o,o' barium, barium, bis 2,4-pentanedionato-o,o'-, t-4, 2,4-pentanedione,ion 1-, barium 2:1, barium, bis 2,4-pentanedionato-o,o'-, hydrate
PubChem CID 5486157
SMILES [Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point >320°C
Physical Form Powder
Quantity 25 g
UN Number UN1564
Solubility Information Insoluble in water.
IUPAC Name barium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 335.56 (Anhydrous)
Chemical Name or Material Barium 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.