missing translation for 'onlineSavingsMsg'
Learn More

Barium 2,4-pentanedionate

Product Code. 11382568
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382568 25 g 25g
11392568 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382568 Supplier Thermo Scientific Alfa Aesar Supplier No. 014613.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12084-29-6 | C10H14BaO4 | 335.55 g/mol

Barium 2,4-pentanedionate hydrate is used in the preparation of radioluminescence of poly ethylene glycolated Eu3+ doped nanophosphors as bioimaging probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12084-29-6
Molecular Formula C10H14BaO4
Molecular Weight (g/mol) 335.55
MDL Number MFCD00013484
InChI Key DJHZYHWLGNJISM-FDGPNNRMSA-L
Synonym barium 2+ ; z-4-oxopent-2-en-2-olate, barium 2,4-pentanedionate, bis pentane-2,4-dionato-o,o' barium, barium, bis 2,4-pentanedionato-o,o'-, t-4, 2,4-pentanedione,ion 1-, barium 2:1, barium, bis 2,4-pentanedionato-o,o'-, hydrate
PubChem CID 5486157
SMILES [Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point >320°C
Physical Form Powder
Quantity 25 g
UN Number UN1564
Solubility Information Insoluble in water.
IUPAC Name barium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 335.56 (Anhydrous)
Chemical Name or Material Barium 2,4-pentanedionate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.